EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Thermostable protease from thermophilic bacteria. I. Thermostability, physiocochemical properties, and amino acid composition.
PMID 5954368