EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Enzymatic formation of cis-homoaconitic acid, an intermediate in lysine biosynthesis in yeast.
PMID 5954805
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
40487
Sodium cis-homoaconitate tribasic, ≥97.0% (HPLC)
C7H5Na3O6