EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Analysis of neutral sugars by gas-liquid chromatography of alditol acetates: application to thyrotropic hormone and other glycoproteins.
PMID 6057635
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91204
Acetic anhydride, for GC derivatization, LiChropur, ≥99.0%
C4H6O3