EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
The chemiluminescence of firefly luciferin. A model for the bioluminescent reaction and identification of the product excited state.
PMID 6064360