EMAIL THIS PAGE TO A FRIEND
Biochemical medicine
Estimation of serum alpha 2-macroglobulin based on the esterolytic activity of bound alpha-chymotrypsin.
PMID 6083782
Abstract
A colorimetric method to estimate alpha 2-macroglobulin (MG) in human serum is described which is based on the capacity of MG:alpha-chymotrypsin complex to hydrolyze N-acetyl L-tyrosine ethyl ester in the presence of excess crude preparation of a trypsin/chymotrypsin inhibitor from redwood seed. Acetyltyrosine formed was measured using Folin's reagent (7). The method was found to be as reliable as, but at least five times more sensitive than the procedures described using trypsin and soybean trypsin inhibitor. MG level is expressed in terms of micrograms of bovine alpha-chymotrypsin bound. Serum from healthy males had a lower value (150.5 +/- 31.9 micrograms/ml, n = 20) than in females (196.8 +/- 40.4, n = 20). No significant difference between the levels in fasting and postprandial conditions was observed.