EMAIL THIS PAGE TO A FRIEND
Doklady Akademii nauk SSSR
[Thermal characteristics of the C--H...O hydrogen bonds formed by nucleic acid base analogs].
PMID 6092025
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
349801
1,3-Dimethyluracil, 99%
C6H8N2O2