EMAIL THIS PAGE TO A FRIEND
Radioisotopes
[Graft polymerization onto poly(methylvinylsiloxane)-bonded silica gel by gamma-ray irradiation].
PMID 6095376
Abstract
Poly(methylvinylsiloxane)-bonded silica gel was synthesized by condensation reaction between silica gel and the hydrolysis product of methylvinyldichlorosilane. Graft polymerization of styrene or indene onto poly(methylvinylsiloxane)-bonded silica gel was carried out by gamma-ray irradiation and compared with that onto silica gel. Homopolymer was found to be formed through a radical mechanism concurrently with an ionic mechanism in the graft polymerization. Probably genuine graft polymers are formed in the graft polymerization onto poly(methylvinylsiloxane)-bonded silica gel, while the product formed in the graft polymerization onto silica gel may be apparent graft polymers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
104914
Dichloromethylvinylsilane, 97%
C3H6Cl2Si