EMAIL THIS PAGE TO A FRIEND
Nature
New class of glutamate agonist structurally related to ibotenic acid.
PMID 6101908
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6816
(±)-AMPA, solid
C7H10N2O4
A9111
(±)-AMPA hydrobromide, ≥98% (HPLC)
C7H10N2O4 · HBr