EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Metronidazole metabolite and Gardnerella vaginalis (Corynebacterium vaginale)
PMID 6112482
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34007
Metronidazole-OH, VETRANAL®, analytical standard
C6H9N3O4