EMAIL THIS PAGE TO A FRIEND
FEBS letters
Regulation of the degradation of 125 I-labeled glutamine synthetase introduced into cultured hepatoma cells by erythrocyte ghost-mediated injection.
PMID 6115767
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
64410
DL-Methionine sulfone, ≥99.0% (NT)
C5H11NO4S
M0876
L-Methionine sulfone
C5H11NO4S