EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
An association between glutamine synthetase activity and adipocyte differentiation in cultured 3T3-L1 cells.
PMID 6117252
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
64410
DL-Methionine sulfone, ≥99.0% (NT)
C5H11NO4S
M0876
L-Methionine sulfone
C5H11NO4S