EMAIL THIS PAGE TO A FRIEND
The American journal of psychiatry
Fluphenazine enanthate and fluphenazine decanoate in the treatment of schizophrenic outpatients: extrapyramidal symptoms and therapeutic effect.
PMID 6120655
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000009 Fluphenazine enantate, European Pharmacopoeia (EP) Reference Standard
C29H38F3N3O2S