EMAIL THIS PAGE TO A FRIEND
Acta tropica
Research on the use of surface active substances in the protection against Schistosoma cercariae.
PMID 6122366
Abstract
Cercariae immobilisation and Schistosoma mansoni adult recovery tests in the mouse show that tensides can greatly reduce the infectivity of cercariae. In the case of two amphoteric products, Laurylbetain and Laurylamidopropylbetain, this action is obtained with very low (0.1-5 ppm) concentrations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43781
(Lauryldimethylammonio)acetate, ≥95% (HPLC)
C16H33NO2