EMAIL THIS PAGE TO A FRIEND
International journal of radiation biology and related studies in physics, chemistry, and medicine
Effect of hyperthermia and ionizing radiation on the erythrocyte membrane.
PMID 6125485
Abstract
Spin-label studies of the effects of hyperthermia on the erythrocyte membrane revealed a decrease in the fluidity of lipids and changes in the state of membrane proteins. The rate of haemolysis in iso-osmotic glycerol solution was increased. Changes of most of the parameters studied when plotted in Arrhenius coordinates revealed a discontinuity (critical hyperthermic transition in the membrane) between 46 and 50 degrees C. Studies of the combined action of ionizing radiation (100 Gy) and hyperthermia (43 degrees C) showed the same direction of changes for (Na-K-Mg)-ATPase activity and spectra of membrane-bound maleimide spin label for both agents, but the additivity of changes depended on the parameter studied.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
253367
4-(2-Iodoacetamido)-TEMPO, free radical
C11H20IN2O2