EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Alpha and beta adrenoceptor blockade in normotensive and deoxycorticosterone (DOC)-hypertensive rats; plasma vasopressin and vasopressin pressor effect.
PMID 6129318