EMAIL THIS PAGE TO A FRIEND
The Medical journal of Australia
Mebhydrolin napadisylate. A possible cause of reversible agranulocytosis and neutropenia.
PMID 6130466
Abstract
Mebhydrolin napadisylate is an antihistamine marketed in Australia as Fabahistin, and as an ingredient of Fabahistin Plus. Since June, 1973, 11 Australian reports associating this drug with either agranulocytosis or neutropenia have been received by the Adverse Drug Reactions Advisory Committee. In seven of these reports, mebhydrolin napadisylate was the only suspected drug. These reactions are probably uncommon, but the association should be borne in mind if evidence of agranulocytosis or neutropenia is found in patients taking the drug.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5279
Mebhydroline 1,5-naphthalenedisulfonate salt, analytical standard
C19H20N2 · 0.5C10H8O6S2