EMAIL THIS PAGE TO A FRIEND
National Cancer Institute monograph
Chemotherapy of human nervous system tumors: influence on cerebrospinal fluid sterols.
PMID 613223
Abstract
The cerebrospinal fluid levels of cholesterol, desmosterol, and the ratio 100 D:C taken from a study of 59 patients are reported. The patients, who had tumors of the nervous system, underwent neurosurgery, radiotherapy, and chemotherapy with 1,3-bis(2-chloroethyl)-1-nitrosourea or 1-(2-chlororethyl)-3-cyclohexyl-1-nitrosourea. The relationships among the levels of these three parameters and the clinical and neuroradiologic evolution are discussed.