EMAIL THIS PAGE TO A FRIEND
Mineral and electrolyte metabolism
PTH secretion in vivo and in vitro. Regulation by calcium and other secretagogues.
PMID 6132328
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
310M-2
Parathyroid Hormone (PTH) (MRQ-31) Mouse Monoclonal Antibody