EMAIL THIS PAGE TO A FRIEND
Neuropatologia polska
[Morphological and histoenzymatic changes in the rat brain after experimental bromfenvinphos (Ipophos-IPO 62) poisoning].
PMID 6135177
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45816
Bromfenvinphos-ethyl, PESTANAL®, analytical standard
C12H14BrCl2O4P