EMAIL THIS PAGE TO A FRIEND
Bollettino chimico farmaceutico
[The pharmacologic activity of 1(2-(p-chloro-alpha-phenylbenzyloxy)ethyl)piperidine (cloperastine)].
PMID 6140927
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2040
Cloperastine hydrochloride, analytical standard
C20H24ClNO · HCl