EMAIL THIS PAGE TO A FRIEND
Bollettino chimico farmaceutico
Simultaneous determination of hydrochloride salts of adiphenine, diphenhydramine, ethyldiphenacetate, drofenine and promazine by ion-pair HPLC.
PMID 6140928