EMAIL THIS PAGE TO A FRIEND
Cutis
"Poppers" or "snappers" dermatitis in homosexual men.
PMID 6148198
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
226637
Butyl nitrite, 95%
C4H9NO2