EMAIL THIS PAGE TO A FRIEND
Archives of toxicology
The action of thallium acetate on spontaneous transmitter release in the rat neuromuscular junction.
PMID 6151386
Abstract
Frequencies and amplitudes of miniature endplate potentials (MEPP's) were recorded from neuromuscular junctions of the rat phrenic nerve-diaphragm preparation in vitro. Superfusion of the preparations with Ringer solution containing thallium acetate (Tlac) gradually increased the frequency of MEPP's by a factor of 10 within 30 min (1 X 10(-3) mol/l Tlac) and 180 min (5 X 10(-4) mol/l Tlac), whereas the amplitude of MEPP's remained unchanged. Frequency of MEPP's fitted a Poisson-distribution which persisted during superfusion with Tl+-Ringer. Sub-MEPP amplitudes remained unaffected by the action of thallium. It is concluded that thallium interferes presynaptically with spontaneous transmitter release.
Related Materials