EMAIL THIS PAGE TO A FRIEND
Annales pharmaceutiques francaises
[Benzonitrile and structural analogs. Action of para-aminobenzonitrile on the central nervous system].
PMID 6152526
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
147753
4-Aminobenzonitrile, 98%
C7H6N2