EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Alanopine and strombine are novel imino acids produced by a dehydrogenase found in the adductor muscle of the oyster, Crassostrea gigas.
PMID 6156653
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M51008
Methyliminodiacetic acid, 99%
C5H9NO4