EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Antiviral and interferon-inducing properties of interferon inducers administered with prostaglandins.
PMID 6158913
Abstract
Prostaglandins enhanced the interferon response and therapeutic activity of interferon inducers in Friend leukemia virus-infected mice. A similar enhancement in interfron responsiveness but not antiviral activity was observed in mice infected with rapidly acute viruses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
205206
2-Amino-5-bromo-6-methyl-4-pyrimidinol, 97%
C5H6BrN3O