EMAIL THIS PAGE TO A FRIEND
Indian journal of experimental biology
Studies on the staining of DNA-aldehyde molecules with Merck's Fuchsin (NB) pulver.
PMID 6162786
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72200
New Fuchsin, for microscopy
C22H23N3 · HCl