EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Bleomycin-induced strand-scission of DNA. Mechanism of deoxyribose cleavage.
PMID 6167580
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
108383
1,1,3,3-Tetramethoxypropane, 99%
C7H16O4