EMAIL THIS PAGE TO A FRIEND
Acta microbiologica Polonica
Effect of microorganisms on the phytotoxicity of herbicides. VI. Modification of venzar activity by bacterial cultures.
PMID 6168180
Abstract
Interactions between two bacterial strains and venzar were compared. It was found that the mechanism of interactions is various and causes the modification of herbicide phytotoxicity. Metabolites of Bacillus subtilis 72 interfered with herbicide by affecting physiological processes in plant tissues and enhancing its inhibitory influence. Arthrobacter sp. 18 strain decreased the phytoinhibitory effect of herbicide due to conjugation with the carrier from venzar.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31112
Lenacil, PESTANAL®, analytical standard
C13H18N2O2