EMAIL THIS PAGE TO A FRIEND
Experientia
Gap junctions in myo-endothelial bridges of rabbit carotid arteries.
PMID 6173257
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
447226
Lanthanum(III) hydroxide, 99.9% trace metals basis
H3LaO3