EMAIL THIS PAGE TO A FRIEND
Immunology letters
The participation of trimethylammonium in the mouse erythrocyte epitope recognized by monoclonal autoantibodies.
PMID 6173316
Abstract
Monoclonal IgM anti-erythrocyte autoantibody produced by a NZB-derived hybridoma has been found to react with trimethylammonium-containing compounds. Such compounds are able to prevent the lysis of bromelain-treated mouse erythrocytes (BrMRBC) by those autoantibodies. Using a column of insolubilized betaine hydrazine (BH) the monoclonal anti-erythrocyte antibody has been specifically retained. Elution of this antibody was accomplished by 0.15 M choline (Ch).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G900
Girard’s reagent T, 99%
C5H14ClN3O