EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Analgesic effect of antagonists of substance P.
PMID 6174127
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0145
[D-Pro2, D-Trp7,9]-Substance P acetate salt, ≥97% (HPLC)
C74H106N20O13S · xC2H4O2