EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Swarm rat chondrosarcoma proteoglycans. Purification of aggregates by zonal centrifugation of preformed cesium sulfate gradients.
PMID 6174523