EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
In vitro histamine release from basophils of asthmatic and atopic individuals in D2O.
PMID 6174620
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B56404
4′-Bromoacetophenone, 98%
C8H7BrO