EMAIL THIS PAGE TO A FRIEND
Carcinogenesis; a comprehensive survey
Quantitative evaluation of dark keratinocytes induced by several promoting and hyperplasiogenic agents: their use as an early morphological indicator of tumor-promoting action.
PMID 6175412
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E45309
Ethyl phenylpropiolate, 98%
C11H10O2