EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Acetylcholinesterase of human erythrocytes and neuromuscular junctions: homologies revealed by monoclonal antibodies.
PMID 6175961