EMAIL THIS PAGE TO A FRIEND
Biochemistry
Coupling factor for photophosphorylation labeled with eosin isothiocyanate: activity, size, and shape in solution.
PMID 6177341
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45245
Eosin 5-isothiocyanate, ≥95.0% (UV)
C21H7Br4NO5S