EMAIL THIS PAGE TO A FRIEND
The Journal of experimental medicine
Direct demonstration of an HLA-DR allotypic determinant on the low molecular weight (beta) subunit using a mouse monoclonal antibody specific for DR3.
PMID 6177816
Abstract
A murine monoclonal antibody directed against a human B cell surface antigen with the characteristics of HLA-DR is described. The antigen detected is tightly linked to HLA and is correlated with the alloantigen HLA-Dw/DR3. Reactivity with a fraction of Dw/DRw6 cells is also observed. The determinant recognized by this antibody has been shown to be present on the smaller molecular weight beta subunit of the HLA-DR antigen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
16-23, Reactivity: Anti-HLA-DR3 and DR52a, 99043001