EMAIL THIS PAGE TO A FRIEND
Mutation research
Mutagens in diesel exhaust particulate. Identification and direct activities of 6-nitrobenzo[a]pyrene, 9-nitroanthracene, 1-nitropyrene and 5h-phenanthro[4,5-bcd]pyran-5-one.
PMID 6178021
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N10209
9-Nitroanthracene, 93%
C14H9NO2
BCR308
9-Nitroanthracene, BCR® certified Reference Material
C14H9NO2