EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Mechanism of [Tyr4]bombesin-induced desensitization in dispersed acini from guinea pig pancreas.
PMID 6181063
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5397
[Tyr4]-Bombesin, ≥97% (HPLC)
C74H108N24O19S