EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Gas-liquid chromatography and enzymatic determination of alanopine and strombine in tissues of marine invertebrates.
PMID 6183991
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M51008
Methyliminodiacetic acid, 99%
C5H9NO4