EMAIL THIS PAGE TO A FRIEND
Der Internist
[Amylase determination using p-nitrophenylmaltosides].
PMID 6185452
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5885
4-Nitrophenyl α-D-maltoside, glycosidase substrate
C18H25NO13