EMAIL THIS PAGE TO A FRIEND
Morphologiai es igazsagugyi orvosi szemle
[Azofloxin--toluidine blue staining].
PMID 6188952
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
210633
Acid Red 1, Dye content 60 %
C18H13N3Na2O8S2
11640
Azophloxine, for microscopy (Hist.)
C18H13N3Na2O8S2
40462
Red 2G, analytical standard
C18H13N3Na2O8S2