EMAIL THIS PAGE TO A FRIEND
Acta veterinaria Scandinavica
Teratogenicity and embryotoxicity of orally administered fenchlorphos in blue foxes.
PMID 6191556
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45485
Fenchlorphos, PESTANAL®, analytical standard
C8H8Cl3O3PS