EMAIL THIS PAGE TO A FRIEND
Neurotoxicology
Consideration of dose levels for delayed neurotoxicity testing in hens: the relationship of neurotoxic dosages to acute LD50 values.
PMID 6192368
Abstract
The oral LD50 of a compound in hens does not reliably predict the neurotoxic dose of that compound. The oral LD50 in rats does not predict either the oral LD50 in hens or the neurotoxic dose in hens, so there is no justification for using the rat value to predict the neurotoxic dosage in hens. A strategy is presented for a stepwise approach to neurotoxicity testing in hens.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N13657 Trichloronate, analytical standard