EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Changes in the urinary metabolites of phylloquinone (vitamin K1) in man following therapeutic anticoagulation with warfarin.
PMID 619911
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19697
Vitamin K1 2,3-epoxide, ≥98.0% (HPLC)
C31H46O3