EMAIL THIS PAGE TO A FRIEND
Journal of immunological methods
The effect of epitope density and antibody affinity on the ELISA as analysed by monoclonal antibodies.
PMID 6205095
Abstract
The antibody titre and the optical density values in an ELISA are influenced by the epitope density of the antigen and the affinity of the antibody tested. This has major implications in the interpretation of ELISA results.
Related Materials