EMAIL THIS PAGE TO A FRIEND
Stain technology
The use of primuline to identify the septum polysaccharide of the fission yeast Schizosaccharomyces pombe.
PMID 6207634
Abstract
Treatment of cells and purified cell walls of the fission yeast Schizosaccharomyces pombe with primuline reveals the septum as a bright fluorescent band. When polysaccharides containing (1----3)-beta-, (1----6)-beta- or (1----3)-alpha-glucosidic linkages are treated with primuline, only those molecules containing chains of (1----3)-beta-glucosyl residues are stained. This implies that (1----3)-beta-glucan is present in the septum of Schiz. pombe as the main constituent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
206865
Primuline, Dye content 50 %
C21H14N3NaO3S3