EMAIL THIS PAGE TO A FRIEND
Life sciences
The antagonism of bombesin in the CNS by substance P analogues.
PMID 6208451
Abstract
The ability of substance P analogues to inhibit the action of bombesin in the CNS was investigated using receptor binding and biological assays. The putative substance P antagonists inhibited binding to central receptors for both substance P and bombesin-like peptides. Spantide, which was the most potent analogue tested, reversed the bombesin induced hypothermia and grooming. Therefore the putative substance P antagonists may also antagonize the actions of bombesin in the CNS.