EMAIL THIS PAGE TO A FRIEND
Philosophical transactions of the Royal Society of London. Series B, Biological sciences
Cloning and characterization of the complementary DNA for the B chain of normal human serum C1q.
PMID 6208566
Abstract
Normal human C1q is a serum glycoprotein of 460 kDa containing 18 polypeptide chains (6A, 6B, 6C) each 226 amino acids long and each containing an N-terminal collagen-like domain and a C-terminal globular domain. Two unusual forms of C1q have been described: a genetically defective form, which has a molecular mass of approximately 160 kDa and is found in the sera of homozygotes for the defect who show a marked susceptibility to immune complex related disease; a fibroblast form, shown to be synthesized and secreted, in vitro, with a molecular mass of about 800 kDa and with chains approximately 16 kDa greater than those of normal C1q. A higher than normal molecular mass form of C1q has also been described in human colostrum and a form of C1q has been claimed to represent one of the types of Fc receptor on guinea-pig macrophages. To initiate studies, at the genomic level, on these various forms of C1q, and to investigate the possible relation between the C1q genes and the procollagen genes, the complementary DNA corresponding to the B chain of normal C1q has been cloned and characterized.